siRNA Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019209.1 | NP_062082.1 | 5 | 365 | ||
GenBank mRNA | BC158690.1 | - | 5 | 365 | 8376316 | |
DY314066.1 | - | 5 | 365 | |||
EV767503.1 | - | 4 | 323 | |||
FQ216026.1 | - | 5 | 358 | |||
FQ216320.1 | - | 5 | 341 | |||
FQ218910.1 | - | 5 | 356 | |||
FQ233169.1 | - | 5 | 342 | |||
FQ234620.1 | - | 5 | 363 | |||
FQ234801.1 | - | 5 | 342 | |||
FQ235247.1 | - | 5 | 342 | |||
Y00350.1 | - | 4 | 321 |

More Information
Other Species Targeted

Gene Ontology Categories:
