siRNA Details
Gene Symbol: Nelfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165901.1 | NP_001159373.1 | 3 | 169 | ||
NM_212548.2 | NP_997713.1 | 3 | 201 | |||
XM_006255921.2 | XP_006255983.1 | 2 | 330 | |||
XM_006255922.2 | XP_006255984.1 | 3 | 379 | |||
GenBank mRNA | BC079427.1 | - | 3 | 201 | 7114415 | |
CO401737.1 | - | 3 | 169 |
