siRNA Details
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017196.3 | NP_058892.1 | 2, 3 | 196 | ||
XM_006256061.2 | XP_006256123.1 | 2, 3 | 291 | |||
XM_006256062.2 | XP_006256124.1 | 3, 4 | 268 | |||
XM_006256063.2 | XP_006256125.1 | 3, 4 | 231 | |||
XM_006256065.2 | XP_006256127.1 | 4, 5 | 346 | |||
GenBank mRNA | AB000818.1 | - | 2, 3 | 194 | ||
BC081822.1 | - | 1, 2 | 764 | |||
D82069.1 | - | 4, 5 | 351 | |||
FM053942.1 | - | 2, 3 | 183 | |||
FQ231499.1 | - | 2, 3 | 194 | |||
U17919.1 | - | 2, 3 | 168 |
