siRNA Details
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017199.1 | NP_058895.1 | 4, 5 | 507 | ||
XM_006229571.2 | XP_006229633.1 | 4, 5 | 563 | |||
GenBank mRNA | BC058482.1 | - | 4, 5 | 420 | 6921620 | |
FQ221421.1 | - | 2, 3 | 123 | |||
FQ221949.1 | - | 2, 3 | 123 | |||
FQ228311.1 | - | 2, 3 | 123 | |||
FQ228501.1 | - | 2, 3 | 123 | |||
Z19087.1 | - | 4, 5 | 507 |
