siRNA Details
Gene Symbol: Dnajb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013907.1 | NP_001013929.1 | 7 | 1075 | ||
XM_006256411.2 | XP_006256473.1 | 7 | 1088 | |||
XM_006256412.2 | XP_006256474.1 | 7 | 1088 | |||
GenBank mRNA | BC090076.1 | - | 7 | 1075 | 7192431 | |
FQ223748.1 | - | 7 | 1055 |
