siRNA Details
Gene Symbol: Lbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017208.2 | NP_058904.2 | 10, 11 | 1169 | ||
GenBank mRNA | BC101906.1 | - | 10, 11 | 1169 | 7933709 | |
FQ219857.1 | - | 3, 4 | 318 |
