siRNA Details
Gene Symbol: Lgals7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022582.2 | NP_072104.2 | 2 | 146 | ||
XM_006228668.2 | XP_006228730.1 | 5 | 404 | |||
XM_006228670.2 | XP_006228732.1 | 4 | 576 | |||
GenBank mRNA | AF036941.1 | - | 2 | 155 | ||
BI278774.1 | - | 2 | 379 |
