siRNA Details
Gene Symbol: Mtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100667.1 | NP_001094137.1 | 1 | 139 | ||
XM_006232612.2 | XP_006232674.1 | 1 | 765 | |||
XM_008761135.1 | XP_008759357.1 | 1 | 777 | |||
GenBank mRNA | BC099090.1 | - | 1 | 50 | ||
BC158632.1 | - | 1 | 139 | 7456627 | ||
FQ216274.1 | - | 1 | 30 |
