siRNA Details
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106449.2 | NP_001099919.1 | 6 | 715 | ||
NM_001270650.1 | NP_001257579.1 | 7 | 789 | |||
NM_001270651.1 | NP_001257580.1 | 7 | 762 | |||
NM_001270652.1 | NP_001257581.1 | 6 | 692 | |||
NM_001270653.1 | NP_001257582.1 | 6 | 719 | |||
XM_006232855.2 | XP_006232917.1 | 7 | 784 | |||
XM_008761282.1 | XP_008759504.1 | 6 | 750 | |||
GenBank mRNA | FM111895.1 | - | 3 | 171 | ||
FQ214903.1 | - | 6 | 652 | |||
FQ215988.1 | - | 7 | 718 | |||
FQ216125.1 | - | 6 | 641 | |||
FQ216236.1 | - | 4 | 386 | |||
FQ216534.1 | - | 6 | 618 | |||
FQ217203.1 | - | 6 | 639 | |||
FQ224853.1 | - | 6 | 640 |
