siRNA Details
Gene Symbol: LOC100912338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232995.2 | XP_006233057.1 | 1 | 129 |
Gene Symbol: Hist2h2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC079138.1 | - | 1 | 90 |
