siRNA Details
Gene Symbol: Myoz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106469.1 | NP_001099939.1 | 5 | 674 | ||
XM_006233264.2 | XP_006233326.1 | 6 | 889 | |||
GenBank mRNA | FQ215232.1 | - | 6 | 784 | ||
FQ215285.1 | - | 6 | 814 | |||
FQ215331.1 | - | 6 | 843 | |||
FQ215936.1 | - | 6 | 784 | |||
FQ216537.1 | - | 7 | 879 | |||
FQ216608.1 | - | 6 | 862 |
