siRNA Details
Gene Symbol: Pebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017236.1 | NP_058932.1 | 4 | 470 | ||
GenBank mRNA | BC063171.1 | - | 4 | 487 | 6888051 | |
FQ211637.1 | - | 4 | 431 | |||
FQ212424.1 | - | 4 | 453 | |||
FQ215504.1 | - | 4 | 481 | |||
FQ224857.1 | - | 4 | 481 | |||
X71873.1 | - | 4 | 445 | |||
X75253.1 | - | 4 | 470 |
