siRNA Details
Gene Symbol: Msh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106477.2 | NP_001099947.2 | 15 | 1944 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106477.2 | NP_001099947.2 | 15 | 1944 |