siRNA Details
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114621.1 | NP_001108093.1 | 9, 10 | 1074 | ||
XM_008762082.1 | XP_008760304.1 | 10, 11 | 2273 | |||
XM_008762083.1 | XP_008760305.1 | 4, 5 | 1005 |
