siRNA Details
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114621.1 | NP_001108093.1 | 19 | 2509 | ||
XM_008762082.1 | XP_008760304.1 | 21 | 3909 | |||
XM_008762083.1 | XP_008760305.1 | 15 | 2641 |
