siRNA Details
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106499.2 | NP_001099969.1 | 5 | 2671 | ||
GenBank mRNA | BC169083.1 | - | 5 | 2671 | 7936869 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106499.2 | NP_001099969.1 | 5 | 2671 | ||
GenBank mRNA | BC169083.1 | - | 5 | 2671 | 7936869 |