siRNA Details
Gene Symbol: RGD1566226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134515.1 | NP_001127987.1 | 6 | 1553 | ||
XM_006234971.2 | XP_006235033.1 | 6 | 2133 | |||
XM_006234972.2 | XP_006235034.1 | 6 | 2133 | |||
XM_008762159.1 | XP_008760381.1 | 6 | 2133 |
