siRNA Details
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017257.2 | NP_058953.1 | 10, 11 | 638 | ||
GenBank mRNA | BC058486.1 | - | 10, 11 | 676 | 6918985 | |
BC097331.1 | - | 10, 11 | 638 | 7389180 | ||
D45250.1 | - | 10, 11 | 688 | |||
FQ227049.1 | - | 10, 11 | 682 | |||
FQ232840.1 | - | 10, 11 | 668 | |||
FQ233763.1 | - | 10, 11 | 664 |
