siRNA Details
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012348.1 | NP_001012348.1 | 3 | 351 | ||
XM_006235015.2 | XP_006235077.1 | 5 | 826 | |||
XM_006235016.2 | XP_006235078.1 | 6 | 2198 | |||
XM_006235017.2 | XP_006235079.1 | 5 | 733 | |||
GenBank mRNA | BC089908.1 | - | 3 | 351 | 7304556 |
