siRNA Details
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106513.1 | NP_001099983.1 | 2 | 495 | ||
XM_008762162.1 | XP_008760384.1 | 2 | 440 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153501, BC106184, AK016755, AK186345, AK154080, and others.
Gene Ontology Categories:
