siRNA Details
Gene Symbol: Ergic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106533.1 | NP_001100003.1 | 11 | 1042 | ||
XM_006235314.1 | XP_006235376.1 | 12 | 1176 | |||
GenBank mRNA | FQ213386.1 | - | 12 | 1028 | ||
FQ230416.1 | - | 11 | 1008 |
