siRNA Details
Gene Symbol: Aar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106535.1 | NP_001100005.1 | 2 | 604 | ||
XM_006235399.2 | XP_006235461.1 | 2 | 547 | |||
XM_008762322.1 | XP_008760544.1 | 2 | 552 | |||
GenBank mRNA | AY724497.1 | - | 2 | 293 | ||
BC161955.1 | - | 1 | 271 | 9027129 |
