siRNA Details
Gene Symbol: Ube2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106542.1 | NP_001100012.1 | 2, 3 | 183 | ||
GenBank mRNA | FQ222472.1 | - | 2, 3 | 174 | ||
FQ234128.1 | - | 2, 3 | 177 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ube2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106542.1 | NP_001100012.1 | 2, 3 | 183 | ||
GenBank mRNA | FQ222472.1 | - | 2, 3 | 174 | ||
FQ234128.1 | - | 2, 3 | 177 |