siRNA Details
Gene Symbol: Nudc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017271.2 | NP_058967.1 | 5 | 657 | ||
GenBank mRNA | BC065581.1 | - | 5 | 643 | 5623848 | |
X82445.1 | - | 5 | 657 |
Gene Symbol: LOC100911422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773882.1 | XP_008772104.1 | 4 | 462 |
