siRNA Details
Gene Symbol: Psx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_231064.2 | XP_231064.2 | 1 | 411 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_231064.2 | XP_231064.2 | 1 | 411 |