siRNA Details
Gene Symbol: RGD1306215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106556.2 | NP_001100026.1 | 6 | 778 | ||
XM_006233588.1 | XP_006233650.1 | 7 | 787 | |||
XM_006233589.1 | XP_006233651.1 | 7 | 781 | |||
GenBank mRNA | BC166714.1 | - | 6 | 778 | 7930772 | |
FQ212908.1 | - | 4 | 948 |
