siRNA Details
Gene Symbol: Lhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106571.1 | NP_001100041.1 | 2 | 241 | ||
XM_006234072.2 | XP_006234134.1 | 2 | 351 | |||
XM_006234073.1 | XP_006234135.1 | 1 | 56 | |||
XM_008761742.1 | XP_008759964.1 | 2 | 366 |
