siRNA Details
Gene Symbol: Psma6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017283.3 | NP_058979.1 | 2 | 197 | ||
XM_006240098.2 | XP_006240160.1 | 2 | 155 | |||
GenBank mRNA | BC062232.1 | - | 2 | 198 | 6888689 | |
D10755.1 | - | 2 | 186 | |||
FQ212808.1 | - | 299 | ||||
FQ217234.1 | - | 2 | 173 | |||
FQ220193.1 | - | 2 | 224 |
