siRNA Details
Gene Symbol: Psmb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017285.1 | NP_058981.1 | 3 | 310 | ||
GenBank mRNA | BC084723.1 | - | 2 | 233 | 7319054 | |
D21800.1 | - | 3 | 310 | |||
FQ211229.1 | - | 3 | 267 | |||
FQ232780.1 | - | 3 | 306 |
