siRNA Details
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106581.1 | NP_001100051.1 | 8 | 872 | ||
XM_006257018.2 | XP_006257080.1 | 7 | 939 | |||
XM_006257019.2 | XP_006257081.1 | 7 | 950 | |||
XM_006257020.2 | XP_006257082.1 | 7 | 941 | |||
GenBank mRNA | BC169078.1 | - | 7 | 886 | 5598223 |
