siRNA Details
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170590.1 | NP_001164061.1 | 8 | 1097 | ||
XM_008773298.1 | XP_008771520.1 | 8 | 1088 | |||
GenBank mRNA | BC169024.1 | - | 1 | 610 | 9029735 | |
CB751131.1 | - | 3 | 277 | |||
CB811034.1 | - | 1 | 154 |
