siRNA Details
Gene Symbol: Pla2g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031598.3 | NP_113786.3 | 5 | 397 | ||
XM_006239146.2 | XP_006239208.1 | 6 | 563 | |||
GenBank mRNA | AY651027.1 | - | 4 | 341 | ||
D00523.1 | - | 5 | 381 | |||
FQ227448.1 | - | 5 | 397 | |||
FQ234749.1 | - | 6 | 495 | |||
M25148.1 | - | 5 | 397 |
