siRNA Details
Gene Symbol: Pla2g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031598.3 | NP_113786.3 | 5 | 443 | ||
XM_006239146.2 | XP_006239208.1 | 6 | 609 | |||
GenBank mRNA | AF365363.1 | - | 4 | 387 | ||
AY651027.1 | - | 4 | 387 | |||
CB772812.1 | - | 1 | 358 | |||
D00523.1 | - | 5 | 427 | |||
FQ220554.1 | - | 3 | 907 | |||
FQ227448.1 | - | 5 | 443 | |||
FQ234749.1 | - | 6 | 541 | |||
M25148.1 | - | 5 | 443 | |||
X74364.1 | - | 3 | 323 |
