siRNA Details
Gene Symbol: Bpgm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199382.1 | NP_955414.1 | 2 | 298 | ||
XM_006236259.2 | XP_006236321.1 | 2 | 412 | |||
GenBank mRNA | AY325190.1 | - | 2 | 207 | ||
BC062240.1 | - | 2 | 298 | 6887677 |
