siRNA Details
Gene Symbol: Mturn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007645.1 | NP_001007646.1 | 2 | 231 | ||
XM_008762880.1 | XP_008761102.1 | 2 | 264 | |||
GenBank mRNA | BC083875.1 | - | 2 | 231 | 7129817 |

More Information
Other Species Targeted

Gene Ontology Categories:
