siRNA Details
Gene Symbol: Rnf181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007647.1 | NP_001007648.1 | 2, 3 | 545 | ||
GenBank mRNA | BC079313.1 | - | 2, 3 | 545 | 7134895 | |
FQ213488.1 | - | 1, 2 | 194 | |||
FQ230795.1 | - | 1, 2 | 144 |
