siRNA Details
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184046.1 | NP_908935.1 | 616 | |||
XM_008763055.1 | XP_008761277.1 | 408 | ||||
GenBank mRNA | AY348867.1 | - | 616 | |||
AY348868.1 | - | 616 |
Gene Symbol: LOC103692165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763036.1 | XP_008761258.1 | 1 | 408 |

More Information
Other Species Targeted

Gene Ontology Categories:
