siRNA Details
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184046.1 | NP_908935.1 | 940 | |||
XM_006236729.2 | XP_006236791.1 | 2968 | ||||
XM_006236731.2 | XP_006236793.1 | 2968 | ||||
XM_006236732.2 | XP_006236794.1 | 2968 | ||||
XM_008763054.1 | XP_008761276.1 | 895 | ||||
XM_008763055.1 | XP_008761277.1 | 732 | ||||
GenBank mRNA | AY348867.1 | - | 940 | |||
AY348868.1 | - | 940 |
Gene Symbol: LOC103692165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763034.1 | XP_008761256.1 | 4 | 2968 | ||
XM_008763035.1 | XP_008761257.1 | 4 | 895 | |||
XM_008763036.1 | XP_008761258.1 | 4 | 732 | |||
XM_008763037.1 | XP_008761259.1 | 4 | 2968 | |||
XM_008763038.1 | XP_008761260.1 | 4 | 2968 |
