siRNA Details
Gene Symbol: Chchd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106608.1 | NP_001100078.1 | 1, 2 | 181 | ||
XM_006236846.2 | XP_006236908.1 | 1, 2 | 188 | |||
GenBank mRNA | FQ219980.1 | - | 1, 2 | 162 |
