siRNA Details
Gene Symbol: Chchd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106608.1 | NP_001100078.1 | 4, 5 | 572 | ||
XM_006236846.2 | XP_006236908.1 | 4, 5 | 579 | |||
GenBank mRNA | FQ219980.1 | - | 4, 5 | 553 |
