siRNA Details
Gene Symbol: Cdca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007648.1 | NP_001007649.1 | 6 | 1204 | ||
XM_006237341.2 | XP_006237403.1 | 6 | 788 | |||
XM_006237342.2 | XP_006237404.1 | 5 | 1221 | |||
XM_008763281.1 | XP_008761503.1 | 6 | 926 | |||
GenBank mRNA | BC079204.1 | - | 6 | 1204 | 7131776 | |
FQ209817.1 | - | 3 | 349 | |||
FQ235136.1 | - | 6 | 742 |
