siRNA Details
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079887.1 | NP_001073356.1 | 4, 5 | 384 | ||
GenBank mRNA | BC086570.1 | - | 4, 5 | 384 | ||
BC127511.1 | - | 4, 5 | 384 | 7376723 | ||
FQ232370.1 | - | 18 |
