siRNA Details
Gene Symbol: Aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106626.1 | NP_001100096.1 | 3 | 440 | ||
XM_006237595.2 | XP_006237657.1 | 5 | 1156 | |||
XM_008763376.1 | XP_008761598.1 | 5 | 1136 | |||
XM_008763377.1 | XP_008761599.1 | 5 | 1157 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009637, NM_001005605, NM_178803, BC025467, AK029011, and others.
Gene Ontology Categories:
