siRNA Details
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106627.3 | NP_001100097.2 | 13, 14 | 1676 | ||
XM_008763378.1 | XP_008761600.1 | 9, 10 | 2113 | |||
GenBank mRNA | BC160907.1 | - | 13, 14 | 1676 | 7455489 | |
CO386521.1 | - | 3, 4 | 413 |
