siRNA Details
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106627.3 | NP_001100097.2 | 14 | 1702 | ||
XM_008763378.1 | XP_008761600.1 | 10 | 2139 | |||
GenBank mRNA | BC160907.1 | - | 14 | 1702 | 7455489 | |
BF546873.1 | - | 1 | 311 | |||
DV722478.1 | - | 2 | 189 |
