siRNA Details
Gene Symbol: Mybl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106632.1 | NP_001100102.1 | 10 | 1563 | ||
XM_006237749.2 | XP_006237811.1 | 10 | 1674 | |||
XM_006237750.2 | XP_006237812.1 | 10 | 2484 | |||
XM_006237751.2 | XP_006237813.1 | 9 | 1569 | |||
XM_006237752.2 | XP_006237814.1 | 10 | 1674 |
