siRNA Details
Gene Symbol: Col15a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100535.1 | NP_001094005.1 | 1475 |
Gene Symbol: LOC100909752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749933.3 | XP_003749981.1 | 8, 9 | 1565 |
