siRNA Details
Gene Symbol: Frem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191757.1 | NP_001178686.1 | 34 | 6255 | ||
XM_006238349.2 | XP_006238411.1 | 35 | 6870 | |||
XM_008763769.1 | XP_008761991.1 | 35 | 6871 | |||
XM_008763770.1 | XP_008761992.1 | 34 | 6356 | |||
XM_008763771.1 | XP_008761993.1 | 31 | 5333 | |||
XM_008763772.1 | XP_008761994.1 | 12 | 1843 |
