siRNA Details
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106670.1 | NP_001100140.1 | 2 | 149 | ||
GenBank mRNA | FQ211202.1 | - | 2 | 228 | ||
FQ211244.1 | - | 2 | 228 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106670.1 | NP_001100140.1 | 2 | 149 | ||
GenBank mRNA | FQ211202.1 | - | 2 | 228 | ||
FQ211244.1 | - | 2 | 228 |