siRNA Details
Gene Symbol: RGD1559786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034132.1 | NP_001029304.1 | 3 | 157 | ||
XM_006238508.2 | XP_006238570.1 | 3 | 273 | |||
XM_006238509.2 | XP_006238571.1 | 2 | 348 | |||
XM_008763867.1 | XP_008762089.1 | 2 | 116 | |||
GenBank mRNA | BC100100.1 | - | 3 | 157 | 7376782 | |
FQ228722.1 | - | 3 | 176 |
